Learn More
Enzo Life Sciences IHC universal negative control reagent (25 ml)
SDP
Catalog No. NC0927745
Encompass_Preferred
IHC universal negative control reagent is specifically designed to work with rabbit, mouse and goat antibodies. The reagent has been titered to work with polymer-based secondary systems in manual and automated IHC platforms. . Formulation: Liquid. Negative control reagent- mixture of purified goat, mouse and rabbit immunoglobulins (IgGs). Applications: IHC. Shipping: Blue Ice Not Frozen. Long Term Storage: +4°C
Catalog No. NC0927745 Supplier Enzo Life Sciences Supplier No. ADI9502310025
IHC universal negative control reagent is specifically designed to work with rabbit, mouse and goat antibodies. The reagent has been titered to work with polymer-based secondary systems in manual and automated IHC platforms. . Formulation: Liquid. Negative control reagent- mixture of purified goat, mouse and rabbit immunoglobulins (IgGs). Applications: IHC. Shipping: Blue Ice Not Frozen. Long Term Storage: +4°C