Learn More
Ihc World LLC IHC WORLD BOUIN'S FLUID 32OZ.
SDP
Catalog No. NC0441300
Encompass
Saturated aqueous picric acid solution. Bouins is commonly used in trichrome staining procedures and with GI tract specimens. Size: 32 oz.
Catalog No. NC0441300 Supplier Ihc World LLC Supplier No. 1175032
Saturated aqueous picric acid solution. Bouins is commonly used in trichrome staining procedures and with GI tract specimens. Size: 32 oz.