Learn More
Pacer Global LLC ihealth Covid Antigen Rapid Test Kits, 2 test per pack, 90 packs per case, 180 tests per case
SDP
Supplier:  Pacer Global LLC MLSICO3000
Encompass_Preferred
iHealth COVID-19 Antigen Rapid Test is the simplest way to detect SARS-CoV-2 nucleocapsid protein antigen. It only takes 4 steps and 15 minutes to complete the test. The test is also non-invasive. You won't need to collect a sample from deep in your nasal cavity to get accurate results.1. Swab-Brush against the inner wall of both nostrils 5 times each in a circular motion with a non-invasive nasal swab. Our nasal swab is soft, highly absorbent, and only needs to be inserted ½ - ¾ inches so you can test yourself comfortably.2. Dip-Insert the swab with the sample into the bottom of the tube and stir it in the fluid 15 times. Squeeze the sides of the tube around the swab as you pull it out. 3. Drip-Put 3 drops of the mixed solution onto the sample port of the COVID-19 test card.4. Wait 15 Minutes-Start the timer. Your results will be ready in 15 minutes.
Catalog No. NC2064145