Learn More
IHEALTH LABS INC iHealth Flu A&B/COVID-19/RSV Rapid Test (1 Pack) - OTC
SDP
Catalog No. NC3959130
Encompass_Preferred
The iHealth Flu A&B/COVID-19/RSV Rapid Test is intended for overthe-counter (OTC) use for the qualitative detection of SARS-CoV-2, influenza A, influenza B, and respiratory syncytial virus (RSV) protein antigens in anterior nasal (nares) swab samples. The test is intended for non-prescription home use by individuals 14 years of age or older testing themselves, or by adults testing individuals 6 months of age or older. It is intended for use in individuals with signs and symptoms of respiratory tract infection within the first six (6) days of symptom onset, when tested at least twice over a three-day period, with a minimum of 48 hours between tests.
Catalog No. NC3959130 Supplier IHEALTH LABS INC Supplier No. CFR1
The iHealth Flu A&B/COVID-19/RSV Rapid Test is intended for overthe-counter (OTC) use for the qualitative detection of SARS-CoV-2, influenza A, influenza B, and respiratory syncytial virus (RSV) protein antigens in anterior nasal (nares) swab samples. The test is intended for non-prescription home use by individuals 14 years of age or older testing themselves, or by adults testing individuals 6 months of age or older. It is intended for use in individuals with signs and symptoms of respiratory tract infection within the first six (6) days of symptom onset, when tested at least twice over a three-day period, with a minimum of 48 hours between tests.