Learn More
IHEALTH LABS INC iHealth Flu A&B/COVID-19/RSV Rapid Test Pro (25 Pack) - POC
SDP
Catalog No. NC3959134
Encompass_Preferred
The iHealth Flu A&B/COVID-19/RSV Rapid Test Pro is a lateral flow immunochromatographic assay intended for the qualitative detection and differentiation of SARS-CoV-2, influenza A, influenza B, and RSV protein antigens directly from anterior nasal swab samples. This test is authorized for use at the Point of Care (POC), i.e., in patient care settings operating under a CLIA Certificate of Waiver, Certificate of Compliance, or Certificate of Accreditation.
Catalog No. NC3959134 Supplier IHEALTH LABS INC Supplier No. CFR25
The iHealth Flu A&B/COVID-19/RSV Rapid Test Pro is a lateral flow immunochromatographic assay intended for the qualitative detection and differentiation of SARS-CoV-2, influenza A, influenza B, and RSV protein antigens directly from anterior nasal swab samples. This test is authorized for use at the Point of Care (POC), i.e., in patient care settings operating under a CLIA Certificate of Waiver, Certificate of Compliance, or Certificate of Accreditation.