Learn More
Medchemexpress LLC HY-P7204 10ug Medchemexpress, IHH, Human CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP720410UG
Encompass_Preferred
Medchemexpress, HY-P7204 10ug IHH, Human CAS: Recombinant Human Indian hedgehog protein is involved in chondrocyte differentiation, proliferation and maturation especially during endochondral ossification. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0582