Learn More
Genscript Corporation IHH Human
Catalog No. 5000059102
Encompass
The Indian Hedgehog protein (IHH) is one of three proteins in the mammalian hedgehog family the others being desert hedgehog (DHH) and Sonic hedgehog (SHH) Hedgehog proteins are important signaling molecules during embryonic development and are highly conserved across species Mouse and human IHH share 100% amino acid identity in the signaling domain while mouse IHH and SHH share 90% amino acid identity in the N-terminal signaling domain IHH mRNA expression is detected in fetal lung gut stomach liver kidney pancreas and strongly in cartilage in growth regions of the developing bone IHH has a specific role in bone growth and differentiation In addition IHH is involved in yolk sac vasculogenesis having a central role in differentiation of epiblast cells into endothelial and red blood cells IHH gene mutations cause the brachydactyly type A1 which is characterized by shortening or malformation of the phalanges and also the acrocapitofemoral dysplasia
Catalog No. 50-000-59102 Supplier Genscript Corporation Supplier No. Z0327710
The Indian Hedgehog protein (IHH) is one of three proteins in the mammalian hedgehog family the others being desert hedgehog (DHH) and Sonic hedgehog (SHH) Hedgehog proteins are important signaling molecules during embryonic development and are highly conserved across species Mouse and human IHH share 100% amino acid identity in the signaling domain while mouse IHH and SHH share 90% amino acid identity in the N-terminal signaling domain IHH mRNA expression is detected in fetal lung gut stomach liver kidney pancreas and strongly in cartilage in growth regions of the developing bone IHH has a specific role in bone growth and differentiation In addition IHH is involved in yolk sac vasculogenesis having a central role in differentiation of epiblast cells into endothelial and red blood cells IHH gene mutations cause the brachydactyly type A1 which is characterized by shortening or malformation of the phalanges and also the acrocapitofemoral dysplasia