Learn More
Medchemexpress LLC Ihmt-PI3Kδ-372 | 2429889-61-0 | 99.7% | C26H23F2N7O2 | 10MG
SDP
Catalog No. 5000260590
Encompass_Preferred
IHMT-PI3Kδ-372 is a potent, selective small-molecule inhibitor of PI3Kδ used in preclinical research. It demonstrates nanomolar potency against PI3Kδ and has documented selectivity, solubility, and storage properties for in vitro and in vivo experiments.
  • Potent PI3Kδ inhibition (IC50 14 nM).
  • High selectivity over other class I PI3Ks (56-83 fold).
  • Minimal inhibition of several CYP enzymes (IC50s > 10 μM for CYP1A2, CYP2B6, CYP2C19, CYP3A4).
  • High purity as analyzed by supplier (≈99.7%).
  • Soluble in DMSO (50 mg/mL) and formulated for in vivo use (≥2.08 mg/mL with specified vehicles).
  • White to off-white solid suitable for analytical and biological assays.
  • Stable when stored as powder at recommended temperatures.
Catalog No. 50-002-60590 Supplier Medchemexpress LLC Supplier No. HY13191010MG
Add to Cart
Add to Cart
IHMT-PI3Kδ-372 is a potent, selective small-molecule inhibitor of PI3Kδ used in preclinical research. It demonstrates nanomolar potency against PI3Kδ and has documented selectivity, solubility, and storage properties for in vitro and in vivo experiments.
  • Potent PI3Kδ inhibition (IC50 14 nM).
  • High selectivity over other class I PI3Ks (56-83 fold).
  • Minimal inhibition of several CYP enzymes (IC50s > 10 μM for CYP1A2, CYP2B6, CYP2C19, CYP3A4).
  • High purity as analyzed by supplier (≈99.7%).
  • Soluble in DMSO (50 mg/mL) and formulated for in vivo use (≥2.08 mg/mL with specified vehicles).
  • White to off-white solid suitable for analytical and biological assays.
  • Stable when stored as powder at recommended temperatures.