Learn More
AdipoGen IHR-NAc (5 mg)
SDP
Supplier:  AdipoGen AGCR13527M005
Encompass_Preferred
Chemical. Formula: C22H16Cl3N3O3. MW: 476.7. Sonic hedgehog (Shh) signaling pathway inhibitor. Potential anticancer compound. Cell membrane permeable Smoothened (Smo) antagonist. Acts on extra- and intracellular exposed Smo. Binds to the seven-transmembrane (7TM) pocket. Blocks hedgehog-induced movement of Smo into the primary cilium, disengages Smo-dependent abrogation of Gli2 and Gli3 proteolytic processing and inhibits Gli activity induced by loss of PTCH1.
Catalog No. 50-205-3915