Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SafeCup™ III 13-F Panel, 25 Cups/Case

Catalog No. 21402414
Change view
Click to view available options
CE Marker:
No
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. CE Marker
21-402-414 No
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-402-414 Supplier SafeCup™ Supplier No. 44415
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rapid qualitative tests for the detection of drugs of abuse in urine

  • Rapid qualitative tests for the detection of drugs of abuse in urine
  • All-in-One Cup, tests for AMP, BAR, BUP, BZO, COC, FEN, MDMA, MET, MOP, MTD, OXY, PCP, THC.
  • 25 Cups per Case, CLIA Waived
  • The Total Collection, Testing, and Transport System
  • Easier to Use, Easier to Read
  • Quick Results in 5 minutes
  • Includes Click Seal in cap for improved leak resistance
  • Temperature Strip Included
  • Store at Room Temperature
  • Adheres to SAMHSA cutoff levels
  • Includes gloves and Security Seals

Specifications

CE Marker No
Clia Complexity Waived
Detectable Analytes Amphetamine (AMP)
Format Drug Cup
Sample Type Urine
Type Drug Cup
Disposable Single-use
Includes Gloves, Security Seals
Quantity 25 Ea.
Test Time 5 Minutes
Form Ready-to-Use (RTU)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.