Learn More
Enzo Life Sciences IIIC3 (500 mg). CAS: 1562-85-2
SDP
Supplier:  Enzo Life Sciences BMLWN1180500
Encompass_Preferred
Inhibits DKK1 of the Wnt pathway. Alternative name: Gallocyanine. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (25mg/ml) or in 100% ethanol (10mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-2375