Learn More
Medchemexpress LLC HY-112897 10mg Medchemexpress, IITZ-01 CAS:1807988-47-1 Purity:>98%
SDP
Catalog No. 501969198
Encompass_Preferred
Medchemexpress, HY-112897 10mg IITZ-01 CAS:1807988-47-1 IITZ-01 is a potent lysosomotropic autophagy inhibitor with single-agent antitumor activity, with an IC50 of 2.62 μM for PI3Kγ. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9198 Supplier Medchemexpress LLC Supplier No. HY11289710MG
Medchemexpress, HY-112897 10mg IITZ-01 CAS:1807988-47-1 IITZ-01 is a potent lysosomotropic autophagy inhibitor with single-agent antitumor activity, with an IC50 of 2.62 μM for PI3Kγ. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.