Learn More
Medchemexpress LLC IK-930 | 2563892-44-2 | C19H19F3N4O2S | 100 MG
SDP
Catalog No. 5000372792
Encompass_Preferred
IK-930 is a potent and orally active TEAD inhibitor with an EC50 value of <0.1 μM. This compound has demonstrated good pharmacokinetic parameters in BALB/c mice, with a Cmax of 1088 ng/mL and an AUC 0-last of 4581 ng*h/mL. It is intended for research use only.
  • Potent and orally active TEAD inhibitor
  • EC50 value of <0.1 μM
  • Demonstrated good pharmacokinetic parameters
  • Undergoing clinical trials for various solid tumors, including malignant pleural mesothelioma and epithelioid hemangioendothelioma
  • For research use only
Catalog No. 50-003-72792 Supplier Medchemexpress LLC Supplier No. HY153585100MG
IK-930 is a potent and orally active TEAD inhibitor with an EC50 value of <0.1 μM. This compound has demonstrated good pharmacokinetic parameters in BALB/c mice, with a Cmax of 1088 ng/mL and an AUC 0-last of 4581 ng*h/mL. It is intended for research use only.
  • Potent and orally active TEAD inhibitor
  • EC50 value of <0.1 μM
  • Demonstrated good pharmacokinetic parameters
  • Undergoing clinical trials for various solid tumors, including malignant pleural mesothelioma and epithelioid hemangioendothelioma
  • For research use only