Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ik-930 | 2563892-44-2 | MFCD34755308 | 99.4% | 424.44 g/mol | C19H19F3N4O2S | 10 MG
SDP

Catalog No. 5000287030
Encompass_Preferred

IK-930 is a small-molecule TEAD inhibitor reported as potent and orally active, with a cellular EC50 <0.1 μM. It is supplied as a solid research reagent for biochemical and cellular studies targeting TEAD-dependent transcription and Hippo pathway biology.

  • Potent, orally active TEAD inhibitor (EC50 <0.1 μM).
  • High purity, 99.39%.
  • Molecular weight 424.44 g/mol.
  • Chemical formula C19H19F3N4O2S.
  • Solid powder form for storage and formulation flexibility.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).
  • CAS number 2563892-44-2 for unambiguous identification.

Catalog No. 50-002-87030 Supplier Medchemexpress LLC Supplier No. HY15358510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

IK-930 is a small-molecule TEAD inhibitor reported as potent and orally active, with a cellular EC50 <0.1 μM. It is supplied as a solid research reagent for biochemical and cellular studies targeting TEAD-dependent transcription and Hippo pathway biology.

  • Potent, orally active TEAD inhibitor (EC50 <0.1 μM).
  • High purity, 99.39%.
  • Molecular weight 424.44 g/mol.
  • Chemical formula C19H19F3N4O2S.
  • Solid powder form for storage and formulation flexibility.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).
  • CAS number 2563892-44-2 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.