Learn More
Apexbio Technology LLC IK-930-50MG
SDP
Catalog No. 5000568700
Encompass_Preferred
IK-930 (CAS No 2563892-44-2) is a small-molecule inhibitor predominantly utilized in cancer research Originating from targeted therapy development it functions by modulating specific signaling pathways involved in tumor proliferation such as the PI3K/AKT/mTOR pathway Its in vitro activity is observed in the nanomolar to low micromolar range which underscores its potency in inhibiting cancer cell growth IK-930 is commonly used in cell models and animal studies to investigate its therapeutic potential and mechanism of action as well as in drug screening processes Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-68700 Supplier Apexbio Technology LLC Supplier No. BA912550
IK-930 (CAS No 2563892-44-2) is a small-molecule inhibitor predominantly utilized in cancer research Originating from targeted therapy development it functions by modulating specific signaling pathways involved in tumor proliferation such as the PI3K/AKT/mTOR pathway Its in vitro activity is observed in the nanomolar to low micromolar range which underscores its potency in inhibiting cancer cell growth IK-930 is commonly used in cell models and animal studies to investigate its therapeutic potential and mechanism of action as well as in drug screening processes Experimental concentrations typically depend on the specific research design