Learn More
Selleck Chemical LLC IK-930-E1483-5MG
SDP
Catalog No. 503134471
Encompass_Preferred
IK-930 is a selective small-molecule inhibitor of TEAD that prevents palmitate binding and thereby disrupts aberrant TEAD-dependent transcription IK-930 demonstrates antitumor activity in mouse xenograft models with Hippo pathway genetic alterations
Catalog No. 50-313-4471 Supplier Selleck Chemical LLC Supplier No. E14835MG
IK-930 is a selective small-molecule inhibitor of TEAD that prevents palmitate binding and thereby disrupts aberrant TEAD-dependent transcription IK-930 demonstrates antitumor activity in mouse xenograft models with Hippo pathway genetic alterations