Learn More
Medchemexpress LLC IK1 inhibitor PA-6 | 500715-03-7 | 99.3% | 492.61 | 50 MG
SDP
Catalog No. 5000437705
Encompass_Preferred
IK1 inhibitor PA-6, a pentamidine analogue, is a selective and potent IK1 (KIR2.x ion-channel-carried inward rectifier current) inhibitor. It has IC50 values of 12-15 nM for human and mouse KIR2.x currents and elevates KIR2.1 protein expression, inducing intracellular KIR2.1 accumulation. This inhibitor has the potential to treat atrial fibrillation and arrhythmia.
  • Selective and potent IK1 inhibitor
  • IC50 values of 12-15 nM for human and mouse KIR2.x currents
  • Elevates KIR2.1 protein expression
  • Induces intracellular KIR2.1 accumulation
  • Potential to treat atrial fibrillation
  • Potential to treat arrhythmia
Catalog No. 50-004-37705 Supplier Medchemexpress LLC Supplier No. HY11254450MG
IK1 inhibitor PA-6, a pentamidine analogue, is a selective and potent IK1 (KIR2.x ion-channel-carried inward rectifier current) inhibitor. It has IC50 values of 12-15 nM for human and mouse KIR2.x currents and elevates KIR2.1 protein expression, inducing intracellular KIR2.1 accumulation. This inhibitor has the potential to treat atrial fibrillation and arrhythmia.
  • Selective and potent IK1 inhibitor
  • IC50 values of 12-15 nM for human and mouse KIR2.x currents
  • Elevates KIR2.1 protein expression
  • Induces intracellular KIR2.1 accumulation
  • Potential to treat atrial fibrillation
  • Potential to treat arrhythmia