Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IK1 inhibitor PA-6 | 500715-03-7 | 99.3% | 492.61 g/mol | C31H32N4O2 | 5 MG
SDP

Catalog No. 5000262735
Encompass_Preferred

IK1 inhibitor PA-6 is a selective and potent inhibitor of inward rectifier potassium (IK1) currents used in electrophysiology and ion-channel research. Supplied as an off-white to light yellow solid, the compound includes detailed solubility and storage specifications for both in vitro and in vivo studies.

  • Selective inhibitor of inward rectifier potassium currents with IC50 ≈ 12-15 nM.
  • High purity (99.29%).
  • Molecular formula C31H32N4O2; molecular weight 492.61 g/mol.
  • Off-white to light yellow solid appearance.
  • Soluble in DMSO at 125 mg/mL; in vivo formulations ≥ 2.08 mg/mL using recommended vehicle protocols.
  • Recommended storage: powder at -20°C (3 years) or 4°C (2 years); solutions at -80°C (2 years) or -20°C (1 year).
  • For research use only; not intended for human or diagnostic use.

Catalog No. 50-002-62735 Supplier Medchemexpress LLC Supplier No. HY1125445MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

IK1 inhibitor PA-6 is a selective and potent inhibitor of inward rectifier potassium (IK1) currents used in electrophysiology and ion-channel research. Supplied as an off-white to light yellow solid, the compound includes detailed solubility and storage specifications for both in vitro and in vivo studies.

  • Selective inhibitor of inward rectifier potassium currents with IC50 ≈ 12-15 nM.
  • High purity (99.29%).
  • Molecular formula C31H32N4O2; molecular weight 492.61 g/mol.
  • Off-white to light yellow solid appearance.
  • Soluble in DMSO at 125 mg/mL; in vivo formulations ≥ 2.08 mg/mL using recommended vehicle protocols.
  • Recommended storage: powder at -20°C (3 years) or 4°C (2 years); solutions at -80°C (2 years) or -20°C (1 year).
  • For research use only; not intended for human or diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.