Learn More
Andwin Scientific Midi MR 1 Digital Magnetic Stirrer
SDP
Catalog No. NC1875562
Encompass_Preferred
Powerful magnetic stirrer without heating function.. Stirring quantity up to 50 liters (H2O). Flat, sturdy stainless steel casing. Non-locking motor. Infinitely variable speed. Digital LED speed display. Timer (0 - 56 min) or continuous operation. Integrated USB and RS 232 interface.
Catalog No. NC1875562 Supplier Andwin Scientific Supplier No. 25002970
Powerful magnetic stirrer without heating function.. Stirring quantity up to 50 liters (H2O). Flat, sturdy stainless steel casing. Non-locking motor. Infinitely variable speed. Digital LED speed display. Timer (0 - 56 min) or continuous operation. Integrated USB and RS 232 interface.