Learn More
Chemglass Life Sciences Stainless Steel Vertical Support Rod
SDP
Catalog No. CG199506
Encompass_Preferred
This stainless steel vertical support rod is 10mm OD and 450mm in length. One end has M10 thread to fit into back right hand corner of hot plate stirrers. Stainless Steel Vertical Support Rod.
Catalog No. CG199506 Supplier Chemglass Life Sciences Supplier No. CG199506
This stainless steel vertical support rod is 10mm OD and 450mm in length. One end has M10 thread to fit into back right hand corner of hot plate stirrers. Stainless Steel Vertical Support Rod.