Learn More
Enzo Life Sciences IκB kinase substrate (biotinylated) (1mg)
SDP
Supplier:  Enzo Life Sciences BMLP1480001
Encompass_Preferred
This peptide sequence, RHDSGLDSMKD, is derived from the area surrounding serine 32 and serine 36 of IκBα, which is phosphorylated by IκB Kinase (IKK). The biotin allows the peptide to be used in IKK kinase assays with streptavidin-bound membranes. Long Term Storage: -20°C.
Catalog No. 50-201-1256