Learn More
Enzo Life Sciences IkappaBalpha (pSer32/36) polyclonal antibody (20 µl)
SDP
Supplier:  Enzo Life Sciences BMLSA4120020
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to a portion of human IκBα phosphorylated at Ser32 and Ser36. Species reactivity: Human, Mouse Porcine. Specificity: Recognizes IκBα phosphorylated at Ser32/36. Applications: WB. Formulation: Liquid. In PBS, pH 7.3, containing glycerol, BSA and sodium azide.
Catalog No. 50-201-1817