Learn More
Sigma Aldrich Fine Chemicals Biosciences IKAR C MAG HS digital IKAM1EA
Catalog No. 502908776
Encompass
New magnetic stirrer with heating and ceramic heating plate which offers excellent chemical resistance.Powerful motor for stirring quantities of up to 15 L (H2O)Simultaneous digital display of target and actual temperatures via LCD displayDirectly connection for a PT 1000 temperature sensor enable a precise temperature control included in delivery)Display of actual temperature value in medium with 0.1 K reSOLUTIONon when using PT 1000 temperature sensor3 operating modes to choose (standard safe adjustment protection)Fixed safety circuit of 550CHot Top indicator hot surface warning to prevent burnsDigital error code displayElevated control panel for protection against leaking liquids
Catalog No. 50-290-8776 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7677271EA
New magnetic stirrer with heating and ceramic heating plate which offers excellent chemical resistance.Powerful motor for stirring quantities of up to 15 L (H2O)Simultaneous digital display of target and actual temperatures via LCD displayDirectly connection for a PT 1000 temperature sensor enable a precise temperature control included in delivery)Display of actual temperature value in medium with 0.1 K reSOLUTIONon when using PT 1000 temperature sensor3 operating modes to choose (standard safe adjustment protection)Fixed safety circuit of 550CHot Top indicator hot surface warning to prevent burnsDigital error code displayElevated control panel for protection against leaking liquids