Learn More
Sigma Aldrich Fine Chemicals Biosciences IKAR C MAG HS hotplate sti1EA
Catalog No. 502909037
Encompass
C-MAG HS 7 IKAMAGNew magnetic stirrer with heating and ceramic heating plate which offers excellent chemical resistance.Powerful motor for stirring quantities of up to 10 L (H2O)Fixed safety circuit of 550CHot Top indicator hot surface warning to prevent burnsExact temperature setting via digital display (LED)Bushing according to DIN 12878 for connecting a contact thermometer e.g. ETS-D5 enables precise temperature controlDigital error code displayElevated control panel for protection against leaking liquids
Catalog No. 50-290-9037 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6717891EA
C-MAG HS 7 IKAMAGNew magnetic stirrer with heating and ceramic heating plate which offers excellent chemical resistance.Powerful motor for stirring quantities of up to 10 L (H2O)Fixed safety circuit of 550CHot Top indicator hot surface warning to prevent burnsExact temperature setting via digital display (LED)Bushing according to DIN 12878 for connecting a contact thermometer e.g. ETS-D5 enables precise temperature controlDigital error code displayElevated control panel for protection against leaking liquids