Learn More
Sigma Aldrich Fine Chemicals Biosciences IKAR C MAG HS hotplate sti1EA
Catalog No. 502908883
Encompass
C-MAG HS 4 IKMAGNew magnetic stirrer with heating and ceramic heating plate which offers excellent chemical resistance.Powerful motor for stirring quantities of up to 5 L (H2O)Fixed safety circuit of 550CHot Top indicator hot surface warning to prevent burnsExact temperature setting via digital display (LED)Digital error code displayElevated control panel for protection against leaking liquids
Catalog No. 50-290-8883 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6717621EA
C-MAG HS 4 IKMAGNew magnetic stirrer with heating and ceramic heating plate which offers excellent chemical resistance.Powerful motor for stirring quantities of up to 5 L (H2O)Fixed safety circuit of 550CHot Top indicator hot surface warning to prevent burnsExact temperature setting via digital display (LED)Digital error code displayElevated control panel for protection against leaking liquids