Learn More
Sigma Aldrich Fine Chemicals Biosciences IKAR EuroStar 60 Control A1EA
Catalog No. 502908669
Encompass
Universal laboratory stirrer designed with a removable wireless controller and a digital TFT display. It automatically adjusts the speed through microprocessor controlled technology within the speed range of 0/30 - 2000 rpm. The stirrer comes equipped with a RS 232 and a USB interface to control and document all parameters. An integrated torque trend display is provided for the measurement of viscosity changes. Safety circuits installed ensures automatic cut-off in an anti-stall or overload conditions. Continuous comparison of shaft speed to desired speed is maintained and variations are adjusted automatically. This guarantees a constant speed even with changes in viscosities of the sample.Infinitely adjustable speedPush-through agitator shaftsOverload protectionShort-term overload operationSlim casingQuiet operationError code display
Catalog No. 50-290-8669 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7669411EA
Universal laboratory stirrer designed with a removable wireless controller and a digital TFT display. It automatically adjusts the speed through microprocessor controlled technology within the speed range of 0/30 - 2000 rpm. The stirrer comes equipped with a RS 232 and a USB interface to control and document all parameters. An integrated torque trend display is provided for the measurement of viscosity changes. Safety circuits installed ensures automatic cut-off in an anti-stall or overload conditions. Continuous comparison of shaft speed to desired speed is maintained and variations are adjusted automatically. This guarantees a constant speed even with changes in viscosities of the sample.Infinitely adjustable speedPush-through agitator shaftsOverload protectionShort-term overload operationSlim casingQuiet operationError code display