Learn More
Sigma Aldrich Fine Chemicals Biosciences IKAR HS 260 basic and cont1EA
Catalog No. 502908875
Encompass
Note Requires AS 260.1 (Z341908) AS 260.2 (Z341916) or AS 260.3 (Z341924) to hold bottles dishes flasks etc.
Catalog No. 50-290-8875 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5618351EA
Note Requires AS 260.1 (Z341908) AS 260.2 (Z341916) or AS 260.3 (Z341924) to hold bottles dishes flasks etc.