Learn More
Sigma Aldrich Fine Chemicals Biosciences IKAR MF10 basic microfine1EA
Catalog No. 502908640
Encompass
IKA(R) MF 10 Basic Microfine Grinder DriveMF 10 basic Microfine grinder drive is a continuously operating universal grinder.MF 10 Basic Microfine Grinder Drive features.nbspPowerful drive.nbspEasy to clean working surface made of stainless steel.nbspTwo different grinding heads can be attached to the drive.nbspHeads are easily changeable.nbspGrinding heads not incl. with deliveryMF 10 Basic Microfine Grinder Drive technical data.nbspProcess type inline.nbspOperating principle cutting/impact.nbspMotor rating input 1000 W.nbspMotor rating output 500 W.nbspSpeed range 3000-6500 rpm.nbspPower-on time ON 120 min.nbspPower-om time OFF 30 min.nbspMilling chamber can be cooled with water no.nbspMill feed can be cooled in milling chamber with dry ice yes.nbspMill feed can be cooled in milling chamber with liquid nitrogen no.nbspDimensions (W x H x D) 320 x 380 x 300 mm.nbspWeight 9.7 kg.nbspPermissible ambient temperature 5-40C.nbspPermissible relative moisture 80%.nbspProtection class accordi
Catalog No. 50-290-8640 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6451681EA
Add to Cart
Add to Cart
Request a Quote
IKA(R) MF 10 Basic Microfine Grinder DriveMF 10 basic Microfine grinder drive is a continuously operating universal grinder.MF 10 Basic Microfine Grinder Drive features.nbspPowerful drive.nbspEasy to clean working surface made of stainless steel.nbspTwo different grinding heads can be attached to the drive.nbspHeads are easily changeable.nbspGrinding heads not incl. with deliveryMF 10 Basic Microfine Grinder Drive technical data.nbspProcess type inline.nbspOperating principle cutting/impact.nbspMotor rating input 1000 W.nbspMotor rating output 500 W.nbspSpeed range 3000-6500 rpm.nbspPower-on time ON 120 min.nbspPower-om time OFF 30 min.nbspMilling chamber can be cooled with water no.nbspMill feed can be cooled in milling chamber with dry ice yes.nbspMill feed can be cooled in milling chamber with liquid nitrogen no.nbspDimensions (W x H x D) 320 x 380 x 300 mm.nbspWeight 9.7 kg.nbspPermissible ambient temperature 5-40C.nbspPermissible relative moisture 80%.nbspProtection class accordi