Learn More
Sigma Aldrich Fine Chemicals Biosciences IKAR RET basic IKAMAGR saf1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z6750591EA
Encompass
IKA(R) RET Basic IKAMAG(TM) Safety ControlWide speed range from 0/50 - 1700 rpmIntegrated temperature controlIncluding PT 1000 temperature sensor (PT 1000.60)Exact temperature and speed setting via digital display even when switched offSet safety temperature limit displayed digitallyHot Top indicator hot surface warning to prevent burnsDigital error code displayWith adjustable safety circuit of heating plate temperature (50 - 380 C)Safety magnetic stirrer with heating suitable for unsupervised operationBushing according to DIN 12878 for connecting a contact thermometer e.g. ETS-D5 enables precise temperature controlHigh level of safety thanks to improved heat control technologyEnclosed assembly (IP 42) guarantees long service lifeVery broad temperature range (RT - 340 C)Extremely fast heating timesElectronic speed controlHigh magnetic adhesionIncluding protection cover H 100
Catalog No. 50-290-9097