Learn More
Sigma Aldrich Fine Chemicals Biosciences IKAR VORTEX 3 vortex shake1EA
Catalog No. 502908547
Encompass
IKA(R) VORTEX 3New vortex shaker suitable for short-time operation (touch function) activated by pressing shaker attachment or continuous operation.Wide speed range infinitely adjustableDifferent applications thanks to 3 interchangeable attachments and 7 inserts (e.g. Eppendorf tubes microtiter plates Erlenmeyer flasks 250 mL etc.) Note please order separatelyAttachments securely click onto appliance in any positionSpecial strap (VG 3.36) ensures easy handling of round/Erlenmeyer flasksSturdy metal zinc die cast casingCompact designShort-time operation activated by pressing attachment (touch function)Stable at high speeds thanks to special feet (silicon base with ultra high vibration damping)Eccentric with ball bearingsSuitable for continuous operation with low self heating thanks to self ventilation of motor
Catalog No. 50-290-8547 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6547601EA
IKA(R) VORTEX 3New vortex shaker suitable for short-time operation (touch function) activated by pressing shaker attachment or continuous operation.Wide speed range infinitely adjustableDifferent applications thanks to 3 interchangeable attachments and 7 inserts (e.g. Eppendorf tubes microtiter plates Erlenmeyer flasks 250 mL etc.) Note please order separatelyAttachments securely click onto appliance in any positionSpecial strap (VG 3.36) ensures easy handling of round/Erlenmeyer flasksSturdy metal zinc die cast casingCompact designShort-time operation activated by pressing attachment (touch function)Stable at high speeds thanks to special feet (silicon base with ultra high vibration damping)Eccentric with ball bearingsSuitable for continuous operation with low self heating thanks to self ventilation of motor