Learn More
Sigma Aldrich Fine Chemicals Biosciences IKAR VXR basic VibraxR orb1EA
Catalog No. 502908764
Encompass
IKA(R) VRX basic Vibrax(R) Orbital ShakersOptoelectronically controlled small shaker with a very wide speed range.Suitable for continuous operationNew design and improved drive systemCircular shaking motionsSlow speeds are well maintainedAttachments are interchangeableExcellent for slow shakingOrbital diameter 4.0AmmMaximum shaking weight (with attachment) 2AkgSpeed range 0 to 2 200Arpm
Catalog No. 50-290-8764 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z4040551EA
IKA(R) VRX basic Vibrax(R) Orbital ShakersOptoelectronically controlled small shaker with a very wide speed range.Suitable for continuous operationNew design and improved drive systemCircular shaking motionsSlow speeds are well maintainedAttachments are interchangeableExcellent for slow shakingOrbital diameter 4.0AmmMaximum shaking weight (with attachment) 2AkgSpeed range 0 to 2 200Arpm