Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology Ikaros Matched Antibody Pair, Human, Mouse, 1 Kit
SDP

Catalog No. 503133173
Encompass

The Ikaros Matched Antibody Pair is designed for high-throughput immunoassay platforms, including ELISA, MSD, Quanterix Simoa, Luminex, and Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF). This pair consists of capture and detection antibodies that bind to non-overlapping epitopes and are supplied in a BSA and azide-free formulation, making them suitable for custom labeling with fluorophores, lanthanides, biotin, or beads.

  • Validated for use in ELISA and ELISA-like assays.
  • Formulated in 1X PBS (pH 7.8) without BSA or azide for compatibility with conjugation.
  • Species reactivity confirmed for human and mouse.
  • Consists of two rabbit monoclonal antibodies (100 μg each).
  • Recommended storage at -20°C in aliquots to minimize freeze/thaw cycles.

Catalog No. 50-313-3173 Supplier Cell Signaling Technology Supplier No. 38302S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Ikaros Matched Antibody Pair is designed for high-throughput immunoassay platforms, including ELISA, MSD, Quanterix Simoa, Luminex, and Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF). This pair consists of capture and detection antibodies that bind to non-overlapping epitopes and are supplied in a BSA and azide-free formulation, making them suitable for custom labeling with fluorophores, lanthanides, biotin, or beads.

  • Validated for use in ELISA and ELISA-like assays.
  • Formulated in 1X PBS (pH 7.8) without BSA or azide for compatibility with conjugation.
  • Species reactivity confirmed for human and mouse.
  • Consists of two rabbit monoclonal antibodies (100 μg each).
  • Recommended storage at -20°C in aliquots to minimize freeze/thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.