Learn More
Cayman Chemical Ikarugamycin, Purity: ≥85%, CAS: 36531-78-9, 2.5 mg
SDP
Catalog No. 502971786
Encompass_Preferred
A macrocyclic antibiotic that exhibits cytotoxic effects in cancer cell lines, inhibiting cell proliferation (IC50 = 221.3 nM in HL-60 cells); 1-4 μM inhibits oxidized LDL-induced accumulation of cholesteryl esters in macrophages; inhibits clathrin-coated pit-mediated endocytosis
Catalog No. 50-297-1786 Supplier Cayman Chemical Supplier No. 153862.5 MG
A macrocyclic antibiotic that exhibits cytotoxic effects in cancer cell lines, inhibiting cell proliferation (IC50 = 221.3 nM in HL-60 cells); 1-4 μM inhibits oxidized LDL-induced accumulation of cholesteryl esters in macrophages; inhibits clathrin-coated pit-mediated endocytosis