Learn More
Medchemexpress LLC IKB beta/NFKBIB Hum 100ug | 100ug
SDP
Catalog No. 5000186329
Encompass_Preferred
IKB beta/NFKBIB protein effectively inhibits NF-kappa-B and prevents NF-kappa-B activation by forming a cytoplasmic complex After cell stimulation the non-phosphorylated form is resynthesized binds to NF-kappa-B aids nuclear transport and protects NF-kappa-B from NFKBIA-dependent inactivation IKB beta/NFKBIB Protein Human (His) is the recombinant human-derived IKB beta/NFKBIB protein expressed by E coli with N-6 His labeled tag
Catalog No. 50-001-86329 Supplier Medchemexpress LLC Supplier No. HYP76986100UG
IKB beta/NFKBIB protein effectively inhibits NF-kappa-B and prevents NF-kappa-B activation by forming a cytoplasmic complex After cell stimulation the non-phosphorylated form is resynthesized binds to NF-kappa-B aids nuclear transport and protects NF-kappa-B from NFKBIA-dependent inactivation IKB beta/NFKBIB Protein Human (His) is the recombinant human-derived IKB beta/NFKBIB protein expressed by E coli with N-6 His labeled tag