Learn More
Aviva Systems Biology Corp IKBKB Antibody - middle region (ARP31988_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31988_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IKBKB. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human IKBKB. Alternate names: FLJ40509, IKK-beta, IKK2, IKKB, MGC131801, NFKBIKBApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54408