Learn More
Aviva Systems Biology Corp IKBKE Antibody - N-terminal region (ARP63182_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63182_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IKBKE. It was validated on Western Blot. . Alternate names: IKK-i, IKKE, IKKI, KIAA0151, MGC125294, MGC125295, MGC125297, IKK-EApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-287-903