Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human IKBKE (NM_014002, 1 a.a. - 716 a.a.) Full-length Recombinant Protein with GST-His tag expressed in Sf9 cells

Catalog No. 89948576 Shop All Abnova Corporation Products
Click to view available options
Quantity:
100 μg

Used for Func, SDS-PAGE

Specifications

Accession Number NM_014002
Concentration 0.055 ug/uL
For Use With (Application) Functional Study, SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 9641
Molecular Weight (g/mol) 110.059kDa
Name IKBKE (Human) Recombinant Protein
Preparation Method Insect cell (Sf9) expression system
Purification Method GST affinity chromatography
Quality Control Testing 2 ug/lane SDS-PAGE Stained with Coomassie Blue
Quantity 100 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing
Regulatory Status RUO
Gene Alias IKK-i/IKKE/IKKI/KIAA0151/MGC125294/MGC125295/MGC125297
Common Name IKBKE
Gene Symbol IKBKE
Biological Activity 42pmol/μg x min
Species Baculovirus
Recombinant Recombinant
Protein Tag GST-His
Expression System Insect cell (Sf9) expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.