Learn More
Aviva Systems Biology Corp IKBKG Antibody - middle region (ARP30005_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30005_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against IKBKG. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human IKBKG. Alternate names: IP, IP1, IP2, FIP3, IPD2, NEMO, FIP-3, Fip3p, AMCBX1, IKK-gammaApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54094