Learn More
Aviva Systems Biology Corp IKBKG Antibody - N-terminal region (ARP30006_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30006_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IKBKG. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human IKBKG. Alternate names: AMCBX1, FIP-3, FIP3, Fip3p, IKK-gamma, IP, IP1, IP2, IPD2, NEMOApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54095