Learn More
Selleck Chemical LLC IKE S8877-100MG
SDP
Catalog No. 5000024778
Encompass_Preferred
IKE(Imidazole ketone erastin) is a potent selective and metabolically stable system xc inhibitor and inducer of ferroptosis
Catalog No. 50-000-24778 Supplier Selleck Chemical LLC Supplier No. S8877100MG
IKE(Imidazole ketone erastin) is a potent selective and metabolically stable system xc inhibitor and inducer of ferroptosis