Learn More
Waters Corp Separation Device, iKey BEH C18, 1.7 μm, 130 Å
Catalog No. 502390574 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The iKey Separation device is a core component of the ionKey/MS system, designed to simplify microflow-LC operations. It eliminates traditional fittings, enhancing user experience for scientists and engineers. Available with a 150 μm internal diameter and 50 mm length, this device makes microflow LC accessible and efficient for analytical applications.
  • Enabling technology for the ionKey/MS system, crucial for advanced analytical workflows.
  • Simplifies microflow-LC by eliminating traditional fittings.
  • Increases accessibility of microflow LC in chromatographic laboratories.
  • Robust performance with BEH C18 hybrid particle, wide pH range (1–10), and high maximum pressure tolerance (10000 psi).
  • Optimized for reversed-phase separations with 1.7 μm particle size, 130 Å pore size, and 18% carbon load.
Catalog No. 50-239-0574 Supplier Waters Corp Supplier No. 186007580
The iKey Separation device is a core component of the ionKey/MS system, designed to simplify microflow-LC operations. It eliminates traditional fittings, enhancing user experience for scientists and engineers. Available with a 150 μm internal diameter and 50 mm length, this device makes microflow LC accessible and efficient for analytical applications.
  • Enabling technology for the ionKey/MS system, crucial for advanced analytical workflows.
  • Simplifies microflow-LC by eliminating traditional fittings.
  • Increases accessibility of microflow LC in chromatographic laboratories.
  • Robust performance with BEH C18 hybrid particle, wide pH range (1–10), and high maximum pressure tolerance (10000 psi).
  • Optimized for reversed-phase separations with 1.7 μm particle size, 130 Å pore size, and 18% carbon load.