Learn More
Waters Corp Waters iKey CSH C18 Separation Device, 1.7 μm, 150 μm × 50 mm
Catalog No. 50727000 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The iKey Separation device is one of the enabling technologies behind the ionKey/MS system. This innovative device eliminates the need for traditional fittings and significantly simplifies the microflow-LC user experience. It is available with an internal diameter of 150 μm and in lengths of 50 mm and 100 mm, making microflow LC accessible to a wider range of chromatographic laboratories.
  • C18 chemistry
  • Reversed phase separation mode
  • Hybrid particle substrate
  • 1 to 10 pH range
  • 10,000 psi (690 Bar) maximum pressure
  • Endcapped
  • Low silanol activity
  • Spherical particle shape
  • 1.7 μm particle size
  • 130 Å pore size
  • Peptide QC tested
  • Nano/Micro system compatibility
  • CSH particle technology
  • L1 USP classification
  • 150 μm inner diameter
  • 50 mm length
  • 15% carbon load
  • 1 unit per package
Catalog No. 50-727-000 Supplier Waters Corp Supplier No. 186007257
Add to Cart
Add to Cart
Request a Quote
The iKey Separation device is one of the enabling technologies behind the ionKey/MS system. This innovative device eliminates the need for traditional fittings and significantly simplifies the microflow-LC user experience. It is available with an internal diameter of 150 μm and in lengths of 50 mm and 100 mm, making microflow LC accessible to a wider range of chromatographic laboratories.
  • C18 chemistry
  • Reversed phase separation mode
  • Hybrid particle substrate
  • 1 to 10 pH range
  • 10,000 psi (690 Bar) maximum pressure
  • Endcapped
  • Low silanol activity
  • Spherical particle shape
  • 1.7 μm particle size
  • 130 Å pore size
  • Peptide QC tested
  • Nano/Micro system compatibility
  • CSH particle technology
  • L1 USP classification
  • 150 μm inner diameter
  • 50 mm length
  • 15% carbon load
  • 1 unit per package