Learn More
Waters Corp Separation Device, 1.8 μm, 300 μm x 50 mm, C18
Catalog No. 502390673 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This separation device is an enabling technology for the ionKey/MS system. This innovative device removes the need for traditional fittings, significantly simplifying the microflow-LC user experience. It makes microflow LC accessible to a wider range of chromatographic laboratories.
  • Eliminates traditional fittings, simplifying the microflow-LC user experience.
  • Makes microflow LC mainstream for chromatographic laboratories.
Catalog No. 50-239-0673 Supplier Waters Corp Supplier No. 186008727
This separation device is an enabling technology for the ionKey/MS system. This innovative device removes the need for traditional fittings, significantly simplifying the microflow-LC user experience. It makes microflow LC accessible to a wider range of chromatographic laboratories.
  • Eliminates traditional fittings, simplifying the microflow-LC user experience.
  • Makes microflow LC mainstream for chromatographic laboratories.