Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp WATERS CORPORATION Separation Device, CSH C18, 1.7 μm, 150 μm, 100 mm

Catalog No. NC0968535 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The iKey Separation device is one of the enabling technologies behind the ionKey/MS system. The innovative iKey eliminates the need for traditional fittings and greatly simplifies the microflow-LC user experience. Available with an internal diameter of 150 μm and in 50 mm and 100 mm lengths, the iKey brings microflow LC into the mainstream of chromatographic laboratories.

  • C18 chemistry
  • Reversed phase separation mode
  • Hybrid particle substrate
  • pH range from 1 pH to 10 pH
  • Maximum pressure of 10000 psi (690 Bar)
  • Particle size of 1.7 μm
  • Pore size of 130 Å
  • Peptide QC tested
  • CSH particle technology
  • Carbon load of 15%
  • Application for peptides

Catalog No. NC0968535 Supplier Waters Corp Supplier No. 186007259
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The iKey Separation device is one of the enabling technologies behind the ionKey/MS system. The innovative iKey eliminates the need for traditional fittings and greatly simplifies the microflow-LC user experience. Available with an internal diameter of 150 μm and in 50 mm and 100 mm lengths, the iKey brings microflow LC into the mainstream of chromatographic laboratories.

  • C18 chemistry
  • Reversed phase separation mode
  • Hybrid particle substrate
  • pH range from 1 pH to 10 pH
  • Maximum pressure of 10000 psi (690 Bar)
  • Particle size of 1.7 μm
  • Pore size of 130 Å
  • Peptide QC tested
  • CSH particle technology
  • Carbon load of 15%
  • Application for peptides

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.