Learn More
Selleck Chemical LLC IKK-16 S2882-50mg
SDP
Catalog No. 503112217
Encompass_Preferred
IKK-16 is a selective I B kinase (IKK) inhibitor for IKK-2 IKK complex and IKK-1 with IC50 of 40 nM 70 nM and 200 nM in cell-free assays respectively IKK-16 also inhibits LRRK2 Ser935 phosphorylation in cells and LRRK2 kinase activity in vitro with IC50 of 50 nM
Catalog No. 50-311-2217 Supplier Selleck Chemical LLC Supplier No. S288250MG
IKK-16 is a selective I B kinase (IKK) inhibitor for IKK-2 IKK complex and IKK-1 with IC50 of 40 nM 70 nM and 200 nM in cell-free assays respectively IKK-16 also inhibits LRRK2 Ser935 phosphorylation in cells and LRRK2 kinase activity in vitro with IC50 of 50 nM