Learn More
Medchemexpress LLC IKK 16 | 873225-46-8 | 99.9% | 483.63 | 200 MG
SDP
Catalog No. 5000348405
Encompass_Preferred
IKK 16 is an orally active IKK inhibitor that targets IKK2, IKK complex, IKK1, and LRRK2. It also acts as a pan-PKD inhibitor and an ABCB1 inhibitor. IKK 16 provides protection against LPS-induced multiple organ dysfunction, aids in restoring renal function and reducing fibrosis in acute kidney injury, and lessens cardiac dysfunction linked to polymicrobial sepsis in type 2 diabetes mellitus models by inhibiting the NF-κB pathway.
  • Orally active IKK inhibitor
  • Targets IKK2, IKK complex, IKK1, and LRRK2
  • Pan-PKD and ABCB1 inhibitor
  • Protects against LPS-induced multiple organ dysfunction
  • Restores renal function and reduces fibrosis in acute kidney injury
  • Lessens cardiac dysfunction in polymicrobial sepsis in T2DM models
Catalog No. 50-003-48405 Supplier Medchemexpress LLC Supplier No. HY13687200MG
IKK 16 is an orally active IKK inhibitor that targets IKK2, IKK complex, IKK1, and LRRK2. It also acts as a pan-PKD inhibitor and an ABCB1 inhibitor. IKK 16 provides protection against LPS-induced multiple organ dysfunction, aids in restoring renal function and reducing fibrosis in acute kidney injury, and lessens cardiac dysfunction linked to polymicrobial sepsis in type 2 diabetes mellitus models by inhibiting the NF-κB pathway.
  • Orally active IKK inhibitor
  • Targets IKK2, IKK complex, IKK1, and LRRK2
  • Pan-PKD and ABCB1 inhibitor
  • Protects against LPS-induced multiple organ dysfunction
  • Restores renal function and reduces fibrosis in acute kidney injury
  • Lessens cardiac dysfunction in polymicrobial sepsis in T2DM models