Learn More
Medchemexpress LLC Ikk 16 (hydrochloride) | 1186195-62-9 | 99.9% | 520.09 | 25 MG
SDP
Catalog No. 5000347944
Encompass_Preferred
IKK 16 hydrochloride is an orally active IKK inhibitor. It also acts as a pan-PKD inhibitor and an ABCB1 inhibitor. This compound protects against LPS-induced multiple organ dysfunction, restores renal function, and alleviates fibrosis in acute kidney injury. It further attenuates cardiac dysfunction associated with polymicrobial sepsis in mice with type 2 diabetes mellitus (T2DM) by inhibiting the NF-κB pathway.
  • Acts as an orally active IKK inhibitor
  • Functions as a pan-PKD inhibitor
  • Inhibits ABCB1
  • Protects against LPS-induced multiple organ dysfunction
  • Restores renal function and alleviates fibrosis in acute kidney injury
  • Attenuates cardiac dysfunction associated with polymicrobial sepsis in mice with type 2 diabetes mellitus
  • Inhibits IκB degradation and TNFα-stimulated expression in HUVEC cells
  • Inhibits phosphorylation in specific LRRK2 cells
Catalog No. 50-003-47944 Supplier Medchemexpress LLC Supplier No. 25MGHY13687A
IKK 16 hydrochloride is an orally active IKK inhibitor. It also acts as a pan-PKD inhibitor and an ABCB1 inhibitor. This compound protects against LPS-induced multiple organ dysfunction, restores renal function, and alleviates fibrosis in acute kidney injury. It further attenuates cardiac dysfunction associated with polymicrobial sepsis in mice with type 2 diabetes mellitus (T2DM) by inhibiting the NF-κB pathway.
  • Acts as an orally active IKK inhibitor
  • Functions as a pan-PKD inhibitor
  • Inhibits ABCB1
  • Protects against LPS-induced multiple organ dysfunction
  • Restores renal function and alleviates fibrosis in acute kidney injury
  • Attenuates cardiac dysfunction associated with polymicrobial sepsis in mice with type 2 diabetes mellitus
  • Inhibits IκB degradation and TNFα-stimulated expression in HUVEC cells
  • Inhibits phosphorylation in specific LRRK2 cells