Learn More
Medchemexpress LLC IKK-16 hydrochloride | 1186195-62-9 | MFCD09971091 | 100.0% | 520.09 g·mol⁻¹ | C28H30ClN5OS | 5 MG
SDP
Catalog No. 5000222029
Encompass_Preferred
IKK-16 hydrochloride is the hydrochloride salt form of a selective IκB kinase (IKK) inhibitor used in research to modulate NF-κB signaling pathways. It is provided as a characterized research reagent for biochemical and cellular studies where inhibition of IKK is required.
  • Selective inhibition of IκB kinase to modulate NF-κB signaling.
  • Suitable for biochemical and cellular assays.
  • High reported purity for reliable experimental results.
  • Hydrochloride salt form improves solubility in common solvents.
  • Available in small quantities for research workflows.
Catalog No. 50-002-22029 Supplier Medchemexpress LLC Supplier No. HY13687A5MG
IKK-16 hydrochloride is the hydrochloride salt form of a selective IκB kinase (IKK) inhibitor used in research to modulate NF-κB signaling pathways. It is provided as a characterized research reagent for biochemical and cellular studies where inhibition of IKK is required.
  • Selective inhibition of IκB kinase to modulate NF-κB signaling.
  • Suitable for biochemical and cellular assays.
  • High reported purity for reliable experimental results.
  • Hydrochloride salt form improves solubility in common solvents.
  • Available in small quantities for research workflows.