Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IKK alpha + IKK beta Antibody (YA349) | 87 kDa | 10 UL
SDP

Catalog No. 5000397882
Encompass_Preferred

IKKβ protein is a serine kinase crucial for the NF-kappa-B signaling pathway, activated by various stimuli including inflammatory cytokines and cellular stresses. As part of the canonical IKK complex, it mediates the conventional NF-kappa-B activation by phosphorylating inhibitors, leading to their degradation and allowing NF-kappa-B to activate gene transcription for immune response, growth control, and protection against apoptosis. IKKβ also phosphorylates other signaling components, regulating canonical IKKs and influencing diverse cellular processes.

  • Rabbit-derived and non-conjugated IgG monoclonal antibody
  • Targets IKK alpha + IKK beta
  • Reactive with human and mouse samples
  • Protein A affinity purified
  • Predicted and observed molecular weight: 87 kDa
  • Applications include WB, ICC/IF, IP
  • Store at -20°C for 1 year, avoid repeated freeze/thaw cycles
  • Shipped with blue ice

Catalog No. 50-003-97882 Supplier Medchemexpress LLC Supplier No. HYP8041410UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

IKKβ protein is a serine kinase crucial for the NF-kappa-B signaling pathway, activated by various stimuli including inflammatory cytokines and cellular stresses. As part of the canonical IKK complex, it mediates the conventional NF-kappa-B activation by phosphorylating inhibitors, leading to their degradation and allowing NF-kappa-B to activate gene transcription for immune response, growth control, and protection against apoptosis. IKKβ also phosphorylates other signaling components, regulating canonical IKKs and influencing diverse cellular processes.

  • Rabbit-derived and non-conjugated IgG monoclonal antibody
  • Targets IKK alpha + IKK beta
  • Reactive with human and mouse samples
  • Protein A affinity purified
  • Predicted and observed molecular weight: 87 kDa
  • Applications include WB, ICC/IF, IP
  • Store at -20°C for 1 year, avoid repeated freeze/thaw cycles
  • Shipped with blue ice

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.