Learn More
Medchemexpress LLC IKK alpha + IKK beta Antibody (YA349) | Protein A affinity purified | Predicted band size: 87 kDa; Observed band size: 87 kDa | 50 UL
SDP
Catalog No. 5000397015
Encompass_Preferred
IKK alpha + IKK beta Antibody (YA349) is a rabbit-derived, non-conjugated IgG monoclonal antibody. It is designed to target IKK alpha + IKK beta. The product has been validated for Western Blot (WB), Immunocytochemistry/Immunofluorescence (ICC/IF), and Immunoprecipitation (IP) applications, and is reactive with human and mouse samples.
  • Rabbit-derived.
  • Non-conjugated IgG monoclonal antibody.
  • Targets IKK alpha + IKK beta.
  • Applicable for WB.
  • Applicable for ICC/IF.
  • Applicable for IP.
  • Reacts with human samples.
  • Reacts with mouse samples.
Catalog No. 50-003-97015 Supplier Medchemexpress LLC Supplier No. HYP8041450UL
IKK alpha + IKK beta Antibody (YA349) is a rabbit-derived, non-conjugated IgG monoclonal antibody. It is designed to target IKK alpha + IKK beta. The product has been validated for Western Blot (WB), Immunocytochemistry/Immunofluorescence (ICC/IF), and Immunoprecipitation (IP) applications, and is reactive with human and mouse samples.
  • Rabbit-derived.
  • Non-conjugated IgG monoclonal antibody.
  • Targets IKK alpha + IKK beta.
  • Applicable for WB.
  • Applicable for ICC/IF.
  • Applicable for IP.
  • Reacts with human samples.
  • Reacts with mouse samples.